Learn More
Gene Tools LLC Endo-Porter PEG, Safe Cytosolic Delivery, Low Toxicity, 1 mL
SDP
Catalog No. NC1501848
Encompass
Endo-Porter PEG is an innovative amphiphilic peptide reagent designed for safe and efficient cytosolic delivery of macromolecules into cultured cells. Its unique endocytosis-mediated mechanism ensures effective cargo escape from endosomes while minimizing toxicity. Unlike other methods, it requires no cargo pre-incubation and works optimally even in 10% serum. This simplifies protocols and offers reduced toxicity, making it a go-to choice for researchers in cellular delivery applications.
Catalog No. NC1501848 Supplier Gene Tools LLC Supplier No. ENDOPORTER (PEG) 1ML
Endo-Porter PEG is an innovative amphiphilic peptide reagent designed for safe and efficient cytosolic delivery of macromolecules into cultured cells. Its unique endocytosis-mediated mechanism ensures effective cargo escape from endosomes while minimizing toxicity. Unlike other methods, it requires no cargo pre-incubation and works optimally even in 10% serum. This simplifies protocols and offers reduced toxicity, making it a go-to choice for researchers in cellular delivery applications.