Learn More
Medchemexpress LLC Endo S2 S. pyogenes 5ug | 5ug
SDP
Catalog No. 5000194967
Encompass_Preferred
Endo S2 secreted by Streptococcus pyogenes deglycosylates the conserved N-linked glycan at Asn297 on IgG Fc to eliminate its effector functions and evade the immune system EndoS2 specifically hydrolyzes diverse glycans on the Fc region of IgG antibodies using a mechanism that relies on its glycoside hydrolase domain and a coevolved carbohydrate-binding module[1][2] Endo S2 Protein S pyogenes (His) is the recombinant Endo S2 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-94967 Supplier Medchemexpress LLC Supplier No. HYP790645UG
Endo S2 secreted by Streptococcus pyogenes deglycosylates the conserved N-linked glycan at Asn297 on IgG Fc to eliminate its effector functions and evade the immune system EndoS2 specifically hydrolyzes diverse glycans on the Fc region of IgG antibodies using a mechanism that relies on its glycoside hydrolase domain and a coevolved carbohydrate-binding module[1][2] Endo S2 Protein S pyogenes (His) is the recombinant Endo S2 protein expressed by E coli with N-6 His labeled tag