Learn More
Med Vet International 5.0mm Cuffed Endotracheal Tube, 10/Box
Catalog No. 501180353
Encompass_Preferred
Murphy type high volume low pressure. Made of thermosensitive clear PVC that softens at body temperature. Radiopaque line to confirm correct tube placement by X-ray. The smooth tip helps to minimize trauma during intubation. Oral or nasal intubation. Magill Curve. Murphy Eye. Sterile. Single Use.
Catalog No. 50-118-0353 Supplier Med Vet International Supplier No. ENDO5.0CX10
Murphy type high volume low pressure. Made of thermosensitive clear PVC that softens at body temperature. Radiopaque line to confirm correct tube placement by X-ray. The smooth tip helps to minimize trauma during intubation. Oral or nasal intubation. Magill Curve. Murphy Eye. Sterile. Single Use.