Learn More
Med Vet International Uncuffed Endotracheal Tube 3.0mm Each
Catalog No. 502830270
Encompass_Preferred
high volume, low-pressure uncuffed ET tubes can be used during pediatric surgical procedures and intensive care. Features include bull-nose tips, split-resistant radiopaque lines, smooth Murphy eyes and kink-resistant thermosensitive tubes.
Catalog No. 50-283-0270 Supplier Med Vet International Supplier No. ENDO3.0UC
high volume, low-pressure uncuffed ET tubes can be used during pediatric surgical procedures and intensive care. Features include bull-nose tips, split-resistant radiopaque lines, smooth Murphy eyes and kink-resistant thermosensitive tubes.