Learn More
New England Biolabs, Inc. Endonuclease VIII – 1000 units
SDP
Catalog No. 50811722
Encompass
Endonuclease VIII from E. coli acts as both an N-glycosylase and an AP-lyase. The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating an apurinic (AP site). The AP-lyase activity cleaves 3' and 5' to the AP site leaving a 5' phosphate and a 3' phosphate. Damaged bases recognized and removed by Endonuclease VIII include urea, 5, 6- dihydroxythymine, thymine glycol, 5-hydroxy-5- methylhydantoin, uracil glycol, 6-hydroxy-5, 6-dihydrothymine and methyltartronylurea. While Endonuclease VIII is similar to Endonuclease III, Endonuclease VIII has and lyase activity while Endonuclease III has only lyase activity.
Catalog No. 50-811-722 Supplier New England Biolabs, Inc. Supplier No. M0299S
Endonuclease VIII from E. coli acts as both an N-glycosylase and an AP-lyase. The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating an apurinic (AP site). The AP-lyase activity cleaves 3' and 5' to the AP site leaving a 5' phosphate and a 3' phosphate. Damaged bases recognized and removed by Endonuclease VIII include urea, 5, 6- dihydroxythymine, thymine glycol, 5-hydroxy-5- methylhydantoin, uracil glycol, 6-hydroxy-5, 6-dihydrothymine and methyltartronylurea. While Endonuclease VIII is similar to Endonuclease III, Endonuclease VIII has and lyase activity while Endonuclease III has only lyase activity.