Learn More
Charles River Laboratories ENDOCHROME-K KET 256 TESTS INC
Catalog No. NC9567565
Encompass
Endochrome-K kit - 256 tests (sensitivity= 50-0.005 EU/ml) 8 X 3.2 ml vials; 2 X 10 ng control standard endotoxin ; 3 X 30 ml LAL reagent water
Catalog No. NC9567565 Supplier Charles River Laboratories Supplier No. R1708K
Endochrome-K kit - 256 tests (sensitivity= 50-0.005 EU/ml) 8 X 3.2 ml vials; 2 X 10 ng control standard endotoxin ; 3 X 30 ml LAL reagent water