Learn More
Enzo Life Sciences EndoG polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905220100
Encompass_Preferred
Host: Rabbit. Isotype: IgG. Immunogen: Synthetic peptide corresponding to 15 amino acids near the N-terminus of human EndoG. The immunogen is located within amino acids 40 - 90 of EndoG. Species reactivity: Human, Mouse, Rat. Applications: ELISA, IHC, IHC (PS), WB. Purity: Affinity chromatography purified via peptide column. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-6946