Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Endoglin/CD105 Antibody Pair [HRP]
SDP

Catalog No. NB009943
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009943 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009943 Supplier R&D Systems™ Supplier No. NBP27945215PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 15.63-1000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Line Novus
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Endoglin/CD105.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Gene ID (Entrez) 2022
Gene Symbol ENG
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 120 ng of recombinant Endoglin/CD105.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Endoglin/CD105 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-Endoglin/CD105 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Endoglin/CD105
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.