Learn More
New England Biolabs, Inc. Endoglycosidase Reaction Buffer Pack – 4 x 1 ml
SDP
Catalog No. 50811533
Encompass
The Endoglycosidase Reaction Buffer Pack contains four buffers: 10X Glycobuffer 2, 10X Glycobuffer 3, 10X Glycoprotein Denaturing Buffer and 10% NP-40, most commonly used in endoglycosidase reactions.
Catalog No. 50-811-533 Supplier New England Biolabs, Inc. Supplier No. B0701S
The Endoglycosidase Reaction Buffer Pack contains four buffers: 10X Glycobuffer 2, 10X Glycobuffer 3, 10X Glycoprotein Denaturing Buffer and 10% NP-40, most commonly used in endoglycosidase reactions.