Learn More
BioVendor, LLC EndoGrade Endotoxin-free Ovalbumin, 1000 mg.
SDP
Supplier:  BioVendor, LLC LET0029
Encompass
EndoGrade Endotoxin-free Ovalbumin, 1000 mg. Endotoxin level < 0.1 EU/mg. Lyophilized. Contains no salts. Purity > 98%. Molecular weight: ~ 45 kDa. Source: Chicken Egg White. Ovalbumin is an extensively used allergen model in immunology and allergology studies. Commercially available ovalbumin is often contaminated with endotoxin, which leads to artefacts and inaccurate results. Endotoxin-free EndoGrade Ovalbumin is of superior quality with a guaranteed endotoxin level (LPS) below 0.1 EU/mg. EndoGrade Ovalbumin is manufactured in Germany by LIONEX GmbH and has been referenced in over 200 scientific publications.
Catalog No. 50-206-9087
Only null left
Add to Cart
Request a Quote