Learn More
MilliporeSigma EndoGRO-VEGF Complete Culture Media Kit
Catalog No. NC1632086
Encompass
The EndoGRO-VEGF Complete Culture Media Kit is a low serum formulation that contains VEGF. It is intended for rapid proliferation of Human Endothelial Cells, HUVEC, aortic endothelial cells & other large vessel endothelial cells.
Catalog No. NC1632086 Supplier MilliporeSigma Supplier No. SCME002S
The EndoGRO-VEGF Complete Culture Media Kit is a low serum formulation that contains VEGF. It is intended for rapid proliferation of Human Endothelial Cells, HUVEC, aortic endothelial cells & other large vessel endothelial cells.