Learn More
Grainger Elga Ultra Filter, 5000 Dalton, Quick Disconnect, 1 ea
Catalog No. 502788948
Encompass_Preferred
The Endoguard 5000 UF membrane is an ultrafiltration module designed for use with pretreated water to remove bacteria, endotoxins, and nucleases. This unit utilizes a quick disconnect connection for straightforward installation and operation in compatible systems.
  • Removes impurities including bacteria, endotoxin, and nucleases
  • Features a 5,000 Dalton molecular weight cutoff for precise filtration
  • Designed for use with pretreated or prefiltered water
  • Equipped with a 5/16 in. quick disconnect connection
  • Operates at a maximum pressure of 60 psi and maximum temperature of 95°F
  • Requires no additional cartridges
  • Recommended replacement interval of 2 years
Catalog No. 50-278-8948 Supplier Grainger Supplier No. 16D251
The Endoguard 5000 UF membrane is an ultrafiltration module designed for use with pretreated water to remove bacteria, endotoxins, and nucleases. This unit utilizes a quick disconnect connection for straightforward installation and operation in compatible systems.
  • Removes impurities including bacteria, endotoxin, and nucleases
  • Features a 5,000 Dalton molecular weight cutoff for precise filtration
  • Designed for use with pretreated or prefiltered water
  • Equipped with a 5/16 in. quick disconnect connection
  • Operates at a maximum pressure of 60 psi and maximum temperature of 95°F
  • Requires no additional cartridges
  • Recommended replacement interval of 2 years