Learn More
World Precision Instrument ENDOHM-12 EVOM™ Electrode for TEER in 12 mm Insert
SDP
Catalog No. NC2241495
Encompass
For TEER measurement of epithelial and endothelial cell cultures
  • The new EndOhm chamber upper mount is made of polycarbonate and unaffected by alcohol
  • The glass chamber is easier to clean and more crack resistant than the prior version
  • Adjustable apical electrode height
  • Crystal clear glass chamber allows visualization of apical electrode positioning
  • New insert holder with 120¿ tri-supports for three leg inserts
  • Three sizes cover a range of well cup sizes from a variety of manufacturers
  • Compatible with the EVOM3 and EVOM ® Manual
Catalog No. NC2241495 Supplier World Precision Instrument Supplier No. EVMEL030102
For TEER measurement of epithelial and endothelial cell cultures
  • The new EndOhm chamber upper mount is made of polycarbonate and unaffected by alcohol
  • The glass chamber is easier to clean and more crack resistant than the prior version
  • Adjustable apical electrode height
  • Crystal clear glass chamber allows visualization of apical electrode positioning
  • New insert holder with 120¿ tri-supports for three leg inserts
  • Three sizes cover a range of well cup sizes from a variety of manufacturers
  • Compatible with the EVOM3 and EVOM ® Manual