Learn More
Surgipro ENDOMETRIAL BIOPSY PIPETTE
Catalog No. NC0308376
Encompass_Preferred
This disposable endometrial sampler/endometrial biopsy pipette has a 3.0mm diameter for maximum specimen yield & it's flexibility ensures virtually no patient discomfort. The pipette's are individually packaged sterile and come in a box of 25 units.
Catalog No. NC0308376 Supplier Surgipro Supplier No. SP0920
Add to Cart
Add to Cart
This disposable endometrial sampler/endometrial biopsy pipette has a 3.0mm diameter for maximum specimen yield & it's flexibility ensures virtually no patient discomfort. The pipette's are individually packaged sterile and come in a box of 25 units.