Learn More
Discovery Life Sciences Endometrial Cancer MSI L DTC
SDP
Catalog No. 5000804084
Encompass
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics
Catalog No. 50-008-04084 Supplier Discovery Life Sciences Supplier No. 320514
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics