Learn More
Medchemexpress LLC HY-P0185 5mg Medchemexpress, Endomorphin 1 CAS:189388-22-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP01855MG
Encompass_Preferred
Medchemexpress, HY-P0185 5mg Endomorphin 1 CAS:189388-22-5 Endomorphin 1, a high affinity, highly selective agonist of the μ-opioid receptor , displays reasonable affinities for kappa 3 binding sites, with K i value between 20 and 30 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4043