Learn More
Medchemexpress LLC Endomorphin 1 acetate | 1276123-71-7 | 99.9% | 670.75 | 25 MG
SDP
Catalog No. 5000426872
Encompass_Preferred
Endomorphin 1 acetate is a highly selective agonist of the μ-opioid receptor (Ki: 1.11 nM). It also shows reasonable affinities for kappa3 binding sites with a Ki value between 20 and 30 nM. This compound possesses antinociceptive properties.
  • Highly selective agonist of the μ-opioid receptor (Ki: 1.11 nM)
  • Displays reasonable affinities for kappa3 binding sites (Ki between 20 and 30 nM)
  • Inhibits Forskolin-stimulated cyclic AMP formation
  • Increases interleukin-8 secretion in Caco-2 cells
  • Inhibits excitatory transmission in adult rat substantia gelatinosa neurons
  • Exhibits antinociceptive properties in mice
  • Alleviates myocardial ischemia/reperfusion injury in rats
Catalog No. 50-004-26872 Supplier Medchemexpress LLC Supplier No. 25MGHYP0185A
Endomorphin 1 acetate is a highly selective agonist of the μ-opioid receptor (Ki: 1.11 nM). It also shows reasonable affinities for kappa3 binding sites with a Ki value between 20 and 30 nM. This compound possesses antinociceptive properties.
  • Highly selective agonist of the μ-opioid receptor (Ki: 1.11 nM)
  • Displays reasonable affinities for kappa3 binding sites (Ki between 20 and 30 nM)
  • Inhibits Forskolin-stimulated cyclic AMP formation
  • Increases interleukin-8 secretion in Caco-2 cells
  • Inhibits excitatory transmission in adult rat substantia gelatinosa neurons
  • Exhibits antinociceptive properties in mice
  • Alleviates myocardial ischemia/reperfusion injury in rats