Learn More
Medchemexpress LLC HY-P0186A 5mg Medchemexpress, Endomorphin 2 (TFA) CAS:1276124-00-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0186A5MG
Encompass_Preferred
Medchemexpress, HY-P0186A 5mg Endomorphin 2 (TFA) CAS:1276124-00-5 Endomorphin 2 TFA, a high affinity, highly selective agonist of the μ-opioid receptor , displays reasonable affinities for kappa 3 binding sites, with K i value between 20 and 30 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4044