Learn More
New England Biolabs, Inc. Endoproteinase LysC – 20 µg
SDP
Catalog No. 501526432
Encompass
Endoproteinase LysC is a serine Endoproteinase which cleaves peptide bonds at the carboxyl side of lysine. LysC is a sequencing grade enzyme and is suitable for proteomics and glycobiology applications.
Catalog No. 50-152-6432 Supplier New England Biolabs, Inc. Supplier No. P8109S
Endoproteinase LysC is a serine Endoproteinase which cleaves peptide bonds at the carboxyl side of lysine. LysC is a sequencing grade enzyme and is suitable for proteomics and glycobiology applications.