Learn More
New England Biolabs, Inc. Endoproteinase AspN – 50 µg
SDP
Catalog No. 50994918
Encompass
Endoproteinase AspN (flavastacin) is a zinc metalloendopeptidase which selectively cleaves protein and peptide bonds N-terminal to aspartic acid residues.
Catalog No. 50-994-918 Supplier New England Biolabs, Inc. Supplier No. P8104S
Endoproteinase AspN (flavastacin) is a zinc metalloendopeptidase which selectively cleaves protein and peptide bonds N-terminal to aspartic acid residues.