Learn More
New England Biolabs, Inc. Endoproteinase GluC – 50 µg
SDP
Catalog No. 50811915
Encompass
Endoproteinase GluC is a serine proteinase which preferentially cleaves peptide bonds C-terminal to glutamic acid residues. It also cleaves at aspartic acid residues at a rate 100-300 times slower than at glutamic acid residues.
Catalog No. 50-811-915 Supplier New England Biolabs, Inc. Supplier No. P8100S
Endoproteinase GluC is a serine proteinase which preferentially cleaves peptide bonds C-terminal to glutamic acid residues. It also cleaves at aspartic acid residues at a rate 100-300 times slower than at glutamic acid residues.