Learn More
Description
- Native endoproteinase Lys-C from Lysobacter enzymogenes
- Serine protease that specifically hydrolyzes amide or ester bonds at the carboxylic side of lysine in peptides and proteins
- Useful for sequence and structural analysis
- Inhibited by aprotinin, DFP, leupeptin, and TLCK
Specifications
Specifications
| pH Range | Optimum 8.5 to 8.8 |
| CAS | 72561-05-8 |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| Quantity | 3 U |
| Solubility | Water |
| Product Type | Endoproteinase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.