Learn More
Worthington Biochemical Corporation Protease, S. aureus, Sequencing Grade, STSEQ, 5x10ug
SDP
Catalog No. NC1752423
Encompass
Protease, S. aureus, Sequencing Grade, STSEQ, 5x10ug. Chromatographically purified according to Drapeau, G, et.al, JBC, 247, 6720 (1972). Supplied as a lyophilized powder. Store at 2-8°C.
Catalog No. NC1752423 Supplier Worthington Biochemical Corporation Supplier No. LS02126
Protease, S. aureus, Sequencing Grade, STSEQ, 5x10ug. Chromatographically purified according to Drapeau, G, et.al, JBC, 247, 6720 (1972). Supplied as a lyophilized powder. Store at 2-8°C.