Learn More
Medchemexpress LLC HY-P1502 1mg Medchemexpress, β-Endorphin, human CAS:61214-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP15021MG
Encompass_Preferred
Medchemexpress, HY-P1502 1mg β-Endorphin, human CAS:61214-51-5 β-Endorphin, human, a prominent endogenous peptide, existing in the hypophysis cerebri and hypothalamus, is an agonist of opioid receptor , with preferred affinity for μ-opioid receptor and δ-opioid receptor ; β-Endorphin, human exhibits antinociception activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4214