Learn More
ACROBiosystems The enzyme hydrolyzes the β-1,4 linkage between the core GlcNAc residues in the Fc-glycan, leaving the innermost GlcNAc intact on the Fc. It is derived from Streptococcus pyogenes and expressed in E. coli. The enzyme contains a His-tag and