Learn More
Charles River Laboratories Endosafe Compendial LAL Cartridges, 0.01, for R&D use only, 10/pk
Catalog No. NC9261714
Encompass
Cartridge; Endosafe-PTS; 0.01 EU/mL sensitivity; Portable, handheld LAL test system
Catalog No. NC9261714 Supplier Charles River Laboratories Supplier No. PTS2001
Cartridge; Endosafe-PTS; 0.01 EU/mL sensitivity; Portable, handheld LAL test system