Learn More
Charles River Laboratories ENDOSAFE PTS CARTRIDGE 0.1 EU
Catalog No. NC9372762
Encompass
Sensitivity: 0.1 EU/ml. This product, PTS201, is unlicensed and is not to be used for pharmaceutical release testing. Please purchase the FDa-licensed product (PTS201F) if you wish to use for release testing.
Catalog No. NC9372762 Supplier Charles River Laboratories Supplier No. PTS201
Sensitivity: 0.1 EU/ml. This product, PTS201, is unlicensed and is not to be used for pharmaceutical release testing. Please purchase the FDa-licensed product (PTS201F) if you wish to use for release testing.