Learn More
Charles River Laboratories Endosafe Compendial LAL Cartridges, 0.05, 10/pk
Catalog No. NC9594798
Encompass
Licensed Cartridge Pack 0.05 EU/ML sensitivity The disposable cartridge contains four channels - two channels with CSE and LAL, which serve as the positive control channels, and two channels with LAL for testing of samples. To perform the test, add 25 ul of sample into all four sample reservoirs. The reader's internal pump moves the samples to the first reagent station for mixing. The pump then moves the samples to the second station for mixing, and lastly into the optical cells of the cartridge to be read kinetically.
Catalog No. NC9594798 Supplier Charles River Laboratories Supplier No. PTS2005F
Licensed Cartridge Pack 0.05 EU/ML sensitivity The disposable cartridge contains four channels - two channels with CSE and LAL, which serve as the positive control channels, and two channels with LAL for testing of samples. To perform the test, add 25 ul of sample into all four sample reservoirs. The reader's internal pump moves the samples to the first reagent station for mixing. The pump then moves the samples to the second station for mixing, and lastly into the optical cells of the cartridge to be read kinetically.