Learn More
Charles River Laboratories Endosafe® Cartridge 10 pack
Catalog No. NC0181821
Encompass
The disposable cartridge contains four channels - two channels with CSE and LaL, which serve as the positive control channels, and two channels with LaL for testing of samples. To perform the test, add 25 al of sample into all four sample reservoirs. The reader's internal pump moves the samples to the first reagent station for mixing. The pump then moves the samples to the second station for mixing, and lastly into the optical cells of the cartridge to be read kinetically. Understand how our FDa-licensed Endosafea-PTSa, c/MCSa, c cartridges are compliant with existing globally harmonized USP/EP guidelines and requirements.
Catalog No. NC0181821 Supplier Charles River Laboratories Supplier No. PTS20005F
The disposable cartridge contains four channels - two channels with CSE and LaL, which serve as the positive control channels, and two channels with LaL for testing of samples. To perform the test, add 25 al of sample into all four sample reservoirs. The reader's internal pump moves the samples to the first reagent station for mixing. The pump then moves the samples to the second station for mixing, and lastly into the optical cells of the cartridge to be read kinetically. Understand how our FDa-licensed Endosafea-PTSa, c/MCSa, c cartridges are compliant with existing globally harmonized USP/EP guidelines and requirements.