Learn More
Charles River Laboratories Gel-Clot LAL Reagent, 0.015 EU/mL, 50-test vial (5.2 mL)
Catalog No. NC9764815
Encompass
The gel-clot assay determines the presence of endotoxins through its clotting response, and is a quick and effective way to detect harmful endotoxins for your injectable pharmaceuticals. Endosafe Rapid LAL Single-Test Vials contain FDA-licensed LAL to provide a simple, self-contained option for routine endotoxin detection in research samples, renal dialysis process water, and dialysate solutions.
Catalog No. NC9764815 Supplier Charles River Laboratories Supplier No. R15015
The gel-clot assay determines the presence of endotoxins through its clotting response, and is a quick and effective way to detect harmful endotoxins for your injectable pharmaceuticals. Endosafe Rapid LAL Single-Test Vials contain FDA-licensed LAL to provide a simple, self-contained option for routine endotoxin detection in research samples, renal dialysis process water, and dialysate solutions.