Learn More
Charles River Laboratories Endosafe Trillium rCR Cartridges
Catalog No. NC3037505
Encompass
Endosafe Trillium rCR Cartridges, 10/pk 10 single packs of Trillium™ rCR cartridges Standard Curve Range: 5 to 0.05 EU/mL
Catalog No. NC3037505 Supplier Charles River Laboratories Supplier No. RCRC3005
Endosafe Trillium rCR Cartridges, 10/pk 10 single packs of Trillium™ rCR cartridges Standard Curve Range: 5 to 0.05 EU/mL