Learn More
Cayman Chemical EndosIdIn 2 10mg
SDP
Catalog No. 502964246
Encompass_Preferred
A cell-permeable inhibitor of exocytosis and endosomal recycling in plant and mammalian cells; binds the EXO70 subunit of the exocyst complex (Kd = 253 μM, EXO70A1); disrupts protein trafficking between the endosome and plasma membrane; enhances trafficking to the vacuole for degradation; inhibits recycling of endocytosed transferrin in HeLa cells
Catalog No. 50-296-4246 Supplier Cayman Chemical Supplier No. 2188810 MG
A cell-permeable inhibitor of exocytosis and endosomal recycling in plant and mammalian cells; binds the EXO70 subunit of the exocyst complex (Kd = 253 μM, EXO70A1); disrupts protein trafficking between the endosome and plasma membrane; enhances trafficking to the vacuole for degradation; inhibits recycling of endocytosed transferrin in HeLa cells