Learn More
Cayman Chemical Endosidin 5, Purity: ≥98%, CAS: 364361-68-2, 10 mg
SDP
Catalog No. 502942797
Encompass_Preferred
An inhibitor of endosomal recycling and dCTPase 1 (IC50 = 1 µM); induces trafficking of endocytosed plasma membrane to the vacuole in Arabidopsis cells; inhibits the recycling of plasma membrane proteins in HeLa cells; inhibits annexin A6-mediated membrane remodeling in HEK293T cell lysates (EC50 = ~3 µM); enhances the delivery of mRNA via LNPs in HEK293T cells in an annexin A6-dependent manner
Catalog No. 50-294-2797 Supplier Cayman Chemical Supplier No. 4132810 MG
An inhibitor of endosomal recycling and dCTPase 1 (IC50 = 1 µM); induces trafficking of endocytosed plasma membrane to the vacuole in Arabidopsis cells; inhibits the recycling of plasma membrane proteins in HeLa cells; inhibits annexin A6-mediated membrane remodeling in HEK293T cell lysates (EC50 = ~3 µM); enhances the delivery of mRNA via LNPs in HEK293T cells in an annexin A6-dependent manner