Learn More
Cell Signaling Technology Endosomal Marker Antibody Sampler Kit, 1 Kit
SDP
Catalog No. NC1312250
Encompass
The Endosomal Marker Antibody Sampler Kit provides an economical means of distinguishing endosomes in the early, late, and recycling phases. Each primary antibody provided is sufficient for performing two western blot experiments.
  • Includes markers for early, late, and recycling endosomal phases
  • Contains primary antibodies for Caveolin-1, Clathrin Heavy Chain, EEA1, Rab5A, Rab7, and Rab11
  • Includes HRP-linked secondary antibodies for convenience
  • Validated for western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation applications
Catalog No. NC1312250 Supplier Cell Signaling Technology Supplier No. 12666T
The Endosomal Marker Antibody Sampler Kit provides an economical means of distinguishing endosomes in the early, late, and recycling phases. Each primary antibody provided is sufficient for performing two western blot experiments.
  • Includes markers for early, late, and recycling endosomal phases
  • Contains primary antibodies for Caveolin-1, Clathrin Heavy Chain, EEA1, Rab5A, Rab7, and Rab11
  • Includes HRP-linked secondary antibodies for convenience
  • Validated for western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation applications