Learn More
Invent Biotechnologies Inc Minute™ Endosome Isolation and Cell Fractionation Kit - Spin Column-Based (20 Preps)
Catalog No. NC1587864
Encompass
Early endosomes, which come mainly from primary endocytic vesicles that fuse with each other, provide the starting point for late endosome maturation. In addition to their roles in normal cell physiology, endocytic processes play a key role in many diseases such as Alzheimer’s disease and inherited lysosomal storage diseases. Traditional methods for isolating endosomes are based on density gradient ultracentrifugation. The protocol requires a large amount of starting material and is tedious and time-consuming. The Minute™ Endosome Isolation and Cell Fractionation Kit provides a spin-column-based novel endosome isolation technology that is rapid and simple and requires a smaller number of cultured cells or milligram amounts of tissues. This kit can precipitate and significantly enrich early endosomes from cultured cells or tissues. More Information: https://inventbiotech.com/products/minute-endosome-isolation-and-cell-fractionation-kit?_pos=1&_sid=fd15de666&_ss=r
Catalog No. NC1587864 Supplier Invent Biotechnologies Inc Supplier No. ED028
Early endosomes, which come mainly from primary endocytic vesicles that fuse with each other, provide the starting point for late endosome maturation. In addition to their roles in normal cell physiology, endocytic processes play a key role in many diseases such as Alzheimer’s disease and inherited lysosomal storage diseases. Traditional methods for isolating endosomes are based on density gradient ultracentrifugation. The protocol requires a large amount of starting material and is tedious and time-consuming. The Minute™ Endosome Isolation and Cell Fractionation Kit provides a spin-column-based novel endosome isolation technology that is rapid and simple and requires a smaller number of cultured cells or milligram amounts of tissues. This kit can precipitate and significantly enrich early endosomes from cultured cells or tissues. More Information: https://inventbiotech.com/products/minute-endosome-isolation-and-cell-fractionation-kit?_pos=1&_sid=fd15de666&_ss=r