Learn More
Medchemexpress LLC HY-P7009 10ug Medchemexpress, Endostatin (P.pastoris-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP700910UG
Encompass_Preferred
Medchemexpress, HY-P7009 10ug Endostatin (P.pastoris-expressed), Human CAS: Recombinant Human Endostatin (P.pastoris-expressed) is an endogenous antiangiogenic peptide with multiple antitumor roles through modulation of various receptors in the plasma membrane, such as suppression of angiogenesis and inhibition of tumor-cell migration and invasion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4261