Learn More
Crescent Chemical Co Inc ENDOSULFAN SULFATE SOLN 1ML
SDP
Catalog No. 5070909
Encompass_Preferred
Endosulfan sulfate Single Component Calibration Standard in Methanol CAS 1031-07-7 Size - 1mL Catalog Number - 7481M.10
Catalog No. 50-709-09 Supplier Crescent Chemical Co Inc Supplier No. CT7481M.10
Endosulfan sulfate Single Component Calibration Standard in Methanol CAS 1031-07-7 Size - 1mL Catalog Number - 7481M.10