Learn More
Cayman Chemical Endosulfn I 50mg
SDP
Catalog No. 502936392
Encompass_Preferred
An organochlorine insecticide; active against bollworms (LD50 = 0.63 mg/g); binds to GABA receptors in rat brain membranes (IC50 = 30 nM); toxic to rats (LD50 = 18 mg/kg)
Catalog No. 50-293-6392 Supplier Cayman Chemical Supplier No. 2425350 MG
An organochlorine insecticide; active against bollworms (LD50 = 0.63 mg/g); binds to GABA receptors in rat brain membranes (IC50 = 30 nM); toxic to rats (LD50 = 18 mg/kg)