Learn More
Cayman Chemical Endosulfan II, Purity: ≥98%, CAS: 33213-65-9, 50 mg
SDP
Catalog No. 502935648
Encompass_Preferred
An organochlorine insecticide; active against a variety of insects including bollworms and tobacco budworms (LD50s = 4.14 and 4.95 mg/g, respectively); binds to GABA receptors (IC50 = 60 nM in rat brain membranes); toxic to rats (LD50 = 240 mg/kg)
Catalog No. 50-293-5648 Supplier Cayman Chemical Supplier No. 2425450 MG
An organochlorine insecticide; active against a variety of insects including bollworms and tobacco budworms (LD50s = 4.14 and 4.95 mg/g, respectively); binds to GABA receptors (IC50 = 60 nM in rat brain membranes); toxic to rats (LD50 = 240 mg/kg)