Learn More
Promocell GmbH PromoCell Endothelial Cell Basal Medium MV 2 (prf) 500 ml, phenol red-free
Catalog No. 50306207
Encompass
Endothelial Cell Growth Medium MV2 is a low-serum (5% V/V) medium developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta.The medium lacks the Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains basic Fibroblast Growth Factor, Insulin-like Growth Factor and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium MV.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 μg / ml || Hydrocortisone | 0.2 μg / ml |
Catalog No. 50-306-207 Supplier Promocell GmbH Supplier No. C22226
Endothelial Cell Growth Medium MV2 is a low-serum (5% V/V) medium developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta.The medium lacks the Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains basic Fibroblast Growth Factor, Insulin-like Growth Factor and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium MV.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 μg / ml || Hydrocortisone | 0.2 μg / ml |