Learn More
Promocell GmbH PromoCell Endothelial Cell Basal Medium 2 (prf) 500 ml, phenol red-free
Catalog No. 50306203
Encompass
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.
Catalog No. 50-306-203 Supplier Promocell GmbH Supplier No. C22216
Add to Cart
Add to Cart
Endothelial Cell Growth Medium 2 is a low-serum (2% V/V) medium that lacks Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains Insulin-like Growth Factor (Long R3 IGF) and Vascular Endothelial Growth Factor (VEGF). Because of its effects on cell metabolism, VEGF may interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.02 ml / ml || Epidermal Growth Factor (recombinant human) | 5 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 10 ng / ml || Insulin-like Growth Factor (Long R3 IGF) | 20 ng / ml || Vascular Endothelial Growth Factor 165 (recombinant human) | 0.5 ng / ml || Ascorbic Acid | 1 µg / ml || Heparin | 22.5 μg / ml || Hydrocortisone | 0.2 µg / ml |For best transfection results we recommend using our heparin-free Endothelial Cell Growth Medium MV2.