Learn More
ATCC Endothelial Cell Growth Kit-VEGF, 1 Frozen Kit
Supplier:  ATCC PCS100041/DEL
Encompass
The Endothelial Cell Growth Kit-VEGF contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030)create a complete, low serum culture environment for human large vessel endothelial cells. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50189649NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-189-649FP