Learn More
ATCC Endothelial Cell Growth Kit-VEGF
Supplier:  ATCC PCS100041
Encompass
The Endothelial Cell Growth Kit-VEGF contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030) create a complete, low serum culture environment for human large vessel endothelial cells. Use of the Endothelial Cell Growth Kit-VEGF will support a faster rate of proliferation because of the presence of several purified human recombinant (rh) growth factors (rh VEGF, rh EGF, rh FGF basic, and rh IGF-1) combined with heparin and hydrocortisone.
Catalog No. 50-238-5081