Learn More
Sigma Aldrich Fine Chemicals Biosciences Endothelial cell growth su5MG
Catalog No. 502900940
Encompass
Endothelial cell growth supplement (ECGS) is an extract of bovine pituitary glands containing growth promoting factors for vascular endothelial cells of mammalian origin. Endothelial cell growth supplement is prepared using a modification of a published method. It is lyophilized from a sterile SOLUTIONon containing NaCl and streptomycin sulfate.
Catalog No. 50-290-0940 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E076015MG
Endothelial cell growth supplement (ECGS) is an extract of bovine pituitary glands containing growth promoting factors for vascular endothelial cells of mammalian origin. Endothelial cell growth supplement is prepared using a modification of a published method. It is lyophilized from a sterile SOLUTIONon containing NaCl and streptomycin sulfate.