Learn More
Sciencell Research Laboratories Endothelial Cell Medium, Complete Kit (includes supplements)
Catalog No. NC9692874
Encompass
Endothelial Cell Medium is a complete medium designed for optimal growth of normal human microvascular endothelial cells in vitro. It is shipped as a kit with FBS, endothelial cell growth supplement, and Pen/Strep solution.
Catalog No. NC9692874 Supplier Sciencell Research Laboratories Supplier No. 1001
Endothelial Cell Medium is a complete medium designed for optimal growth of normal human microvascular endothelial cells in vitro. It is shipped as a kit with FBS, endothelial cell growth supplement, and Pen/Strep solution.