Learn More
Cayman Chemical Endothelial Lipase human Blo
SDP
Catalog No. 502886990
Encompass_Preferred
Peptide Sequence amino acids 19-32 (AGSPVPFGPEGRLE) · To be used in conjunction with Cayman’s endothelial lipase polyclonal antibody (Catalog No. 100030) to block protein-antibody complex formation during analysis for the endothelial lipase.
Catalog No. 50-288-6990 Supplier Cayman Chemical Supplier No. 100041111 EA
Peptide Sequence amino acids 19-32 (AGSPVPFGPEGRLE) · To be used in conjunction with Cayman’s endothelial lipase polyclonal antibody (Catalog No. 100030) to block protein-antibody complex formation during analysis for the endothelial lipase.