Learn More
TARGETMOL CHEMICALS INC ENDOTHELIAL LIPASE INHIB 5MG
SDP
Catalog No. 502581552
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. purity: 97%
Catalog No. 50-258-1552 Supplier TARGETMOL CHEMICALS INC Supplier No. T111995MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. purity: 97%