Learn More
Medchemexpress LLC Endothelial lipase inhibitor-1 | 1466427-02-0 | 98.0% | 406.43 | 1 MG
SDP
Catalog No. 5000439380
Encompass_Preferred
Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. It is for research use only and not sold to patients.
  • Potent endothelial lipase inhibitor with an IC50 of 49 nM.
  • For research use only.
Catalog No. 50-004-39380 Supplier Medchemexpress LLC Supplier No. HY1129111MG
Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. It is for research use only and not sold to patients.
  • Potent endothelial lipase inhibitor with an IC50 of 49 nM.
  • For research use only.