Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Endothelial lipase inhibitor-1 | 1466427-02-0 | 98.0% | 406.43 | 1 ML
SDP

Catalog No. 5000439588
Encompass_Preferred

Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. This compound is for research use only and not sold to patients.

  • Potent endothelial lipase inhibitor with an IC50 of 49 nM.
  • Molecular weight: 406.43.
  • Purity: 98.02%.
  • Soluble in DMSO at 25 mg/mL.
  • For research use only.

Catalog No. 50-004-39588 Supplier Medchemexpress LLC Supplier No. HY11291110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endothelial lipase inhibitor-1 is a potent endothelial lipase inhibitor with an IC50 of 49 nM. This compound is for research use only and not sold to patients.

  • Potent endothelial lipase inhibitor with an IC50 of 49 nM.
  • Molecular weight: 406.43.
  • Purity: 98.02%.
  • Soluble in DMSO at 25 mg/mL.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.