Learn More
Apexbio Technology LLC Endothelin-1 (1-15), amide, human 1mg
SDP
Catalog No. 5000093122
Encompass_Preferred
Endothelin-1 1-15 amide human is a peptide fragment derived from the N-terminus of Endothelin-1 which targets endothelin receptor subtypes ETA and ETB It is designed to modulate vascular tone through activation of G protein-coupled receptor signaling thereby influencing blood pressure regulation and vascular function Endothelin-1 1-15 amide human exerts its biological activity primarily through receptor activation and signal transduction mechanisms involving endothelin receptors Based on these pharmacological properties Endothelin-1 1-15 amide human holds research potential in the investigation of receptor-ligand interactions receptor selectivity and the pathophysiology of hypertension cancer diabetes pain modulation and cerebral vasospasm
Catalog No. 50-000-93122 Supplier Apexbio Technology LLC Supplier No. A11111
Endothelin-1 1-15 amide human is a peptide fragment derived from the N-terminus of Endothelin-1 which targets endothelin receptor subtypes ETA and ETB It is designed to modulate vascular tone through activation of G protein-coupled receptor signaling thereby influencing blood pressure regulation and vascular function Endothelin-1 1-15 amide human exerts its biological activity primarily through receptor activation and signal transduction mechanisms involving endothelin receptors Based on these pharmacological properties Endothelin-1 1-15 amide human holds research potential in the investigation of receptor-ligand interactions receptor selectivity and the pathophysiology of hypertension cancer diabetes pain modulation and cerebral vasospasm